ContactUsLinkComponent
Restek Scott/Air Liquide Air Std Replacement for p/n 34512 14L, 5% ea of Carbon Monoxide, Carbon Dioxide and Nitrogen, 4% of Methane and Hydrogen, 2.5% Oxygen (Note change in oxygen percentage) (incl.Dangerous Goods Fee) 1 * 1 ST
Catalog # RESTRT-34543
Supplier:  Restek
Restek Scott/Air Liquide Air Std Replacement for p/n 34512 14L, 5% ea of Carbon Monoxide, Carbon Dioxide and Nitrogen, 4% of Methane and Hydrogen, 2.5% Oxygen (Note change in oxygen percentage) (incl.Dangerous Goods Fee) 1 * 1 ST
Catalog # RESTRT-34543
Supplier:  Restek
Frequently Bought Together