ContactUsLinkComponent
ATP serves as a marker for cell viability as it is present in all metabolically active cells. Lumino Cell Viability Glow Assay is a bioluminescent assay to detect and/or quantify ATP and thus measures the viable cells. 1 * 1.000 Assays
Catalog # GENO786-1358
Supplier:  G-Biosciences
ATP serves as a marker for cell viability as it is present in all metabolically active cells. Lumino Cell Viability Glow Assay is a bioluminescent assay to detect and/or quantify ATP and thus measures the viable cells. 1 * 1.000 Assays
Catalog # GENO786-1358
Supplier:  G-Biosciences
Frequently Bought Together