Affinity purification of butyrylcholinesterase (BuChE). BuChE can be purified by batch adsorption on immobilized Procainamide follow ed by ion-exchange chromatography on DEAE-Sepharose column. Immobilized Procainamide can also be used to purify acet 1 * 100 mL
Catalog # GENO786-1320
Supplier: G-Biosciences
Frequently Bought Together