ContactUsLinkComponent
Toxicity screening is a powerful technique that allows scientists to determine the effect of potentially harmful substances on livin g cells. In this inquiry-based lab, students will plan and implement a toxicity screening experiment using insect cel 1 * 1 KIT
Catalog # EDVO1002
Supplier:  EDVOTEK
Toxicity screening is a powerful technique that allows scientists to determine the effect of potentially harmful substances on livin g cells. In this inquiry-based lab, students will plan and implement a toxicity screening experiment using insect cel 1 * 1 KIT
Catalog # EDVO1002
Supplier:  EDVOTEK
Frequently Bought Together