ContactUsLinkComponent
MiniLab a taste of genetics - Extraction, PCR and electrophoresis reagents. Includes DNA extraction solution, Taq polymerase, primers, restriction enzyme, GreenGel cups, running buffer and accessoires for ten groups of students. 1 * 1 KIT
Catalog # EMBIM6010
Supplier:  EMBITEC
MiniLab a taste of genetics - Extraction, PCR and electrophoresis reagents. Includes DNA extraction solution, Taq polymerase, primers, restriction enzyme, GreenGel cups, running buffer and accessoires for ten groups of students. 1 * 1 KIT
Catalog # EMBIM6010
Supplier:  EMBITEC
Frequently Bought Together