ContactUsLinkComponent
MiniLab colourful dye electrophoresis. Includes colourful dye samples (9), GreenGel cups (10), TBE concentrate (100ml), 0,65ml micro centrifuge tubes (100 tubes), micropipette tips (120 tips) for ten groups of students. Teacher and student guides. 1 * 1 KIT
Catalog # EMBIM3007
Supplier:  EMBITEC
MiniLab colourful dye electrophoresis. Includes colourful dye samples (9), GreenGel cups (10), TBE concentrate (100ml), 0,65ml micro centrifuge tubes (100 tubes), micropipette tips (120 tips) for ten groups of students. Teacher and student guides. 1 * 1 KIT
Catalog # EMBIM3007
Supplier:  EMBITEC
Frequently Bought Together