ContactUsLinkComponent
MiniLab foodborne outbreak investigation. Includes DNA Samples (11), GreenGel cups (10), TBE concentrate (100ml), 0,65ml microcentri fuge tubes (120 tubes), micropipette tips (120 tips) for ten groups of students. Teacher and student guides. 1 * 1 KIT
Catalog # EMBIM3006
Supplier:  EMBITEC
MiniLab foodborne outbreak investigation. Includes DNA Samples (11), GreenGel cups (10), TBE concentrate (100ml), 0,65ml microcentri fuge tubes (120 tubes), micropipette tips (120 tips) for ten groups of students. Teacher and student guides. 1 * 1 KIT
Catalog # EMBIM3006
Supplier:  EMBITEC
Frequently Bought Together