ContactUsLinkComponent
MiniOne electrophoresis system. Includes electrophoresis unit, blue light illumination, gel casting system, photo hood with amber fi lter, 42V power supply. Comes with free 2-20ml pipette and a validation kit. 1 * 1 items
  EMBIM1000
 :  EMBITEC
MiniOne electrophoresis system. Includes electrophoresis unit, blue light illumination, gel casting system, photo hood with amber fi lter, 42V power supply. Comes with free 2-20ml pipette and a validation kit. 1 * 1 items
  EMBIM1000
 :  EMBITEC
Frequently Bought Together