ContactUsLinkComponent
NECTIN4 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human Nectin-4 in cell cultu re supernates, cell lysates, serum, plasma (heparin, EDTA) and urine. 1 * 1 KIT
Catalog # ABNOKA5175
Supplier:  Abnova
NECTIN4 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human Nectin-4 in cell cultu re supernates, cell lysates, serum, plasma (heparin, EDTA) and urine. 1 * 1 KIT
Catalog # ABNOKA5175
Supplier:  Abnova
Frequently Bought Together