ContactUsLinkComponent
Sartopure GF Plus MidiCap Retention Rate: 0.65 µm Glass fibre fleeces depth filter capsule with defined particle retention. Technical data Retention Rate: 0.65 µm Filtration area: 0.12 m² Construction: 53 Connectors: Inlet 1 1/2 inch Tri - Clamp Outl 1 * 4 items
  SARP5555305P9-SS-A
 :  Sartorius
Sartopure GF Plus MidiCap Retention Rate: 0.65 µm Glass fibre fleeces depth filter capsule with defined particle retention. Technical data Retention Rate: 0.65 µm Filtration area: 0.12 m² Construction: 53 Connectors: Inlet 1 1/2 inch Tri - Clamp Outl 1 * 4 items
  SARP5555305P9-SS-A
 :  Sartorius
Frequently Bought Together