ContactUsLinkComponent
ZH 102 Spark plug holder for 12 spark plugs with thread dimensions of 19 mm max. 1 * 1 items
Catalog # BAND383
ZH 102 Spark plug holder for 12 spark plugs with thread dimensions of 19 mm max. 1 * 1 items
Catalog # BAND383
Frequently Bought Together