Orders LinkContactUsLinkComponent
Colour coders for cryovials, CryoColorCode, Nunc™
Colour coders for cryovials, CryoColorCode, Nunc™
Grey
Catalog # 479-2486
   Low Manufacturing Impact
Colour coders for cryovials, CryoColorCode, Nunc™
Catalog # 479-2486
   Low Manufacturing Impact
Colour coders for cryovials, CryoColorCode, Nunc™
Colour coders for cryovials, CryoColorCode, Nunc™
Grey
Price
27,60 €
Log in for availability
1 * 500 items
Product Details & Documents
Colour coded inserts for the cap with inscription area.
Notch to remove tube or CryoColorCodes.
Specifications
  • Colour:
    Grey
  • Seq:
    13
  • Cat. No.:
    479-2486
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Environmentally Preferable:
    Low Manufacturing Impact