ContactUsLinkComponent
BALANCE MS304TS/M00 ANALYT 320G X 0.1MG 1 * 1 items
Catalog # METB30133555
Supplier:  Mettler - Toledo
BALANCE MS304TS/M00 ANALYT 320G X 0.1MG 1 * 1 items
Catalog # METB30133555
Supplier:  Mettler - Toledo
Frequently Bought Together