Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Stericlin® sterilisation wrap interleaved, alternately stacked sheets, non-woven 52g/m² blue and green, 140x140cm, DIN EN ISO 11607 1 * 100 items
Catalog # 129-1350
Supplier:
VP Medical Packaging
Expand image
Stericlin® sterilisation wrap interleaved, alternately stacked sheets, non-woven 52g/m² blue and green, 140x140cm, DIN EN ISO 11607 1 * 100 items
Catalog # 129-1350
Supplier:
VP Medical Packaging
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals