ContactUsLinkComponent
Stericlin® barcode-labels, on roll, for thermal transfer printers, paper, double self-adhesive (sandwich), with STEAM indicator, lab el size 70x35mm, roll width 73mm x core 40mm, pack of 10 rolls of 1000 labels 1 * 10.000 items
Catalog # VEPA3FSKS675155
Stericlin® barcode-labels, on roll, for thermal transfer printers, paper, double self-adhesive (sandwich), with STEAM indicator, lab el size 70x35mm, roll width 73mm x core 40mm, pack of 10 rolls of 1000 labels 1 * 10.000 items
Catalog # VEPA3FSKS675155
Frequently Bought Together