ContactUsLinkComponent
Stericlin® sterilisation wrap sheet, non-woven 52g/m² green, 75x75cm, DIN EN ISO 11607 1 * 200 items
Catalog # 129-1383
Stericlin® sterilisation wrap sheet, non-woven 52g/m² green, 75x75cm, DIN EN ISO 11607 1 * 200 items
Catalog # 129-1383
Frequently Bought Together