ContactUsLinkComponent
Column Protection System Guard Column Holder CC for CC 8 x 3 mm and 8 x 4 mm guard columns consisting of: - 1 guard column holder with steel capillary 0.12 mm ID special PEEK ferrule and connector to main column - 2 double open ended wrenches 12 and 1 * 1 SET
Catalog # MANA718964
Supplier:  MACHEREY-NAGEL
Column Protection System Guard Column Holder CC for CC 8 x 3 mm and 8 x 4 mm guard columns consisting of: - 1 guard column holder with steel capillary 0.12 mm ID special PEEK ferrule and connector to main column - 2 double open ended wrenches 12 and 1 * 1 SET
Catalog # MANA718964
Supplier:  MACHEREY-NAGEL
Frequently Bought Together