ContactUsLinkComponent
Pipette Tips LQR LTS 1000µL F 960/10 High throughput liqud handling platform tips, LTS 1ML 10 racks / 96 tips filter 1 * 960 items
Catalog # RAIN30296782
Supplier:  RAININ
Pipette Tips LQR LTS 1000µL F 960/10 High throughput liqud handling platform tips, LTS 1ML 10 racks / 96 tips filter 1 * 960 items
Catalog # RAIN30296782
Supplier:  RAININ
Frequently Bought Together