Agilent J+W DB-5HT Intuvo GC column module, 30 m, 0.25 mm, 0.10 µm 1 * 1 items
HEWL122-5731-INT
: Agilent
Agilent J+W DB-5HT Intuvo GC column module, 30 m, 0.25 mm, 0.10 µm 1 * 1 items
HEWL122-5731-INT
: Agilent
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection