Agilent J+W DB-17 Intuvo GC column module, 30 m, 0.32 mm, 0.50 µm 1 * 1 items
HEWL123-1733-INT
: Agilent
Agilent J+W DB-17 Intuvo GC column module, 30 m, 0.32 mm, 0.50 µm 1 * 1 items
HEWL123-1733-INT
: Agilent
Frequently Bought Together
Acetone ≥99%, TECHNICAL