Stirrer heating mantles WHM. F. round bottom flasks 6000ml, with integrated power controller, temperature range 450C,. Magnetic stirrer: 50-750 rpm, 800w, power supply 230V, 1 * 1 items
Catalog # WITGDH.WHM12038
Supplier: witeg Labortechnik
Frequently Bought Together