ContactUsLinkComponent
EREG (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human epiregulin in cell cultur e supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5094
Supplier:  Abnova
EREG (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human epiregulin in cell cultur e supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5094
Supplier:  Abnova
Frequently Bought Together