Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Stericlin® sterilisation wrap sheet, SSMMS 43g/m², one side light blue, one side violet, 90x90cm, DIN EN ISO 11607 1 * 200 items
Catalog # 129-1361
Supplier:
VP Medical Packaging
Certificates
Expand image
Stericlin® sterilisation wrap sheet, SSMMS 43g/m², one side light blue, one side violet, 90x90cm, DIN EN ISO 11607 1 * 200 items
Catalog # 129-1361
Supplier:
VP Medical Packaging
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
CoC_VEPA3FSMS360112_1252173
CoC_VEPA3FSMS360112_1278545
CoC_VEPA3FSMS360112_1290490
CoC_VEPA3FSMS360112_1295077
CoC_VEPA3FSMS360112_1311970
More
Documentation
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection