ContactUsLinkComponent
CryoSette Frozen Tissue Storage Containers. Base made of acetal, cap made of high-density polyethylene. 2.5 ml container with wide- mouth opening. 1 * 50 items
Catalog # SIMPM956O
Supplier:  Simport Scientific
CryoSette Frozen Tissue Storage Containers. Base made of acetal, cap made of high-density polyethylene. 2.5 ml container with wide- mouth opening. 1 * 50 items
Catalog # SIMPM956O
Supplier:  Simport Scientific
Frequently Bought Together