TNC (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of human TNC in cell culture supernates, serum, plasma (heparin, EDTA) and urine. 1 * 1 KIT
Catalog # ABNOKA5082
Supplier: Abnova
Frequently Bought Together