ContactUsLinkComponent
Autom. burettes, Pellet, 25 ml:0,05 DIN-AS, with ST-stopcock, with lateral stopcock, ST-glass- plug, 2ltr. bottle, amber glass, conf.certified, without aspirator bulb 1 * 1 items
  WITG3.124.256
Autom. burettes, Pellet, 25 ml:0,05 DIN-AS, with ST-stopcock, with lateral stopcock, ST-glass- plug, 2ltr. bottle, amber glass, conf.certified, without aspirator bulb 1 * 1 items
  WITG3.124.256
Frequently Bought Together