ContactUsLinkComponent
Hydrometer DIN 12794/BS 718, L50, 1,05-1,10:0,005g/cm3, ref Temp 20C, without therm, max 335mm long, government calibrated, with DKD-Certificate with 3 test points 1 * 1 items
Catalog # AMARH806218/DKD3
Supplier:  Amarell
Hydrometer DIN 12794/BS 718, L50, 1,05-1,10:0,005g/cm3, ref Temp 20C, without therm, max 335mm long, government calibrated, with DKD-Certificate with 3 test points 1 * 1 items
Catalog # AMARH806218/DKD3
Supplier:  Amarell
Frequently Bought Together