ContactUsLinkComponent
Allopregnanolone ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of allopregnanolone in extracted s erum, plasma, or dried fecal samples, or in diluted urine and tissue culture media samples. 1 * 1 KIT
Catalog # ABNOKA5050
Supplier:  Abnova
Allopregnanolone ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of allopregnanolone in extracted s erum, plasma, or dried fecal samples, or in diluted urine and tissue culture media samples. 1 * 1 KIT
Catalog # ABNOKA5050
Supplier:  Abnova
Frequently Bought Together