Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

Human Recombinant CNTF (from E. coli)

Vial, 20 µG
Catalog # SHBT100-59-20UG
Human Recombinant CNTF (from <i>E. coli</i>)
Catalog # SHBT100-59-20UG
CAS Number:  
Human Recombinant CNTF (from <i>E. coli</i>)
Human Recombinant CNTF (from E. coli)

Vial, 20 µG

CAS Number:  
Price
190,00 €
Log in for availability
1 * 20 µG
Product Details & Documents

Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrowth, and neurotransmitter synthesis.CNTF also plays an important protective role during nervous system injury.

  • High quality
  • Low endotoxin
  • Affordable

 : For research use only.

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    20 µG
  • Protease-free:
    N
  • Purity:
    ≥95%
  • Source:
    E. coli
  • Molecular weight:
    22,9 kDa
  • Nuclease-free:
    N
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Protein function:
    Cytokine
  • Gene ID:
    P26441
  • Formulation:
    10 mM Sodium phosphate, pH 7,5
  • Endotoxin-free:
    N
  • Tested applications:
    Bioassay
  • Species:
    Human
  • Protein/peptide name:
    CNTF
  • Endotoxin level:
    Low
  • Grade:
    RUO
  • Sequence:
    MAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM
  • Biological activity:
    Bioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤325 ng/ml, Bioactivity ≥3,1×10³ units/mg
  • Shipping temperature:
    RT
  • Cat. No.:
    SHBT100-59-20UG
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Storage conditions:
    –20 °C
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Animal-free:
    N

Specifications