Orders LinkContactUsLinkComponent
Accessories for REVOLVE
Accessories for REVOLVE
Stage insert, Hemocytometer
Catalog # ECHLHLDR0006
Supplier:   DISCOVER ECHO
Accessories for REVOLVE
Catalog # ECHLHLDR0006
Supplier:  DISCOVER ECHO
Accessories for REVOLVE
Stage insert, Hemocytometer
Price
135,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Stage insert, Hemocytometer
  • Seq:
    08
  • Cat. No.:
    ECHLHLDR0006
  • Observation method:
    Brightfield and phase contrast
  • Output:
    USB
  • Electrical:
    167 - 240 VA (50/60 Hz)
  • Camera:
    79 MP colour
  • Display:
    12,9” iPad Pro® Retina® display, 2732×2048 (264 pixels/inch)
  • Sensor:
    CMOS