Orders LinkContactUsLinkComponent
Fluorite Objectives for Revolve Microscope
Apochromat oil phase, 0,15 mm
Catalog # ECHLJMAG0602
Supplier:   DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Catalog # ECHLJMAG0602
Supplier:  DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Apochromat oil phase, 0,15 mm
Price
8 210,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    60×
  • Description:
    Apochromat oil phase
  • Seq:
    03
  • Working distance:
    0,15 mm