Orders LinkContactUsLinkComponent
Fluorite Objectives for Revolve Microscope
Fluorite Objectives for Revolve Microscope
Fluorite objective, 2,1 mm
Catalog # 630-3057
Supplier:   DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Catalog # 630-3057
Supplier:  DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Fluorite objective, 2,1 mm
Price
1 250,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Documents
Specifications
  • Magnification:
    20×
  • Description:
    Fluorite objective
  • Seq:
    16
  • Working distance:
    2,1 mm