Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Achromat phase objective, 10 mm
Catalog # ECHLJMAG0105
Supplier:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Catalog # ECHLJMAG0105
Supplier:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Achromat phase objective, 10 mm
Price
394,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Documents
Specifications
  • Magnification:
    10×
  • Description:
    Achromat phase objective
  • Seq:
    08
  • Working distance:
    10 mm