ContactUsLinkComponent
Hydrorometer DIN12791/BS 718, S50, 1,05-1,10:0,0020g/cm3, ref temp 20C, without therm, max 190mm long, government calibrated with DKD certificate 3 test points 1 * 1 items
Catalog # AMARH811118DKD3
Supplier:  Amarell
Hydrorometer DIN12791/BS 718, S50, 1,05-1,10:0,0020g/cm3, ref temp 20C, without therm, max 190mm long, government calibrated with DKD certificate 3 test points 1 * 1 items
Catalog # AMARH811118DKD3
Supplier:  Amarell
Frequently Bought Together