Orders LinkContactUsLinkComponent
Fluorite Objectives for Revolve Microscope
Fluorite Objectives for Revolve Microscope
Fluorite ELWD phase objective, 1,5 - 2,2 mm
Catalog # 630-3064
Supplier:   DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Catalog # 630-3064
Supplier:  DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Fluorite ELWD phase objective, 1,5 - 2,2 mm
Price
6 420,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Documents
Specifications
  • Magnification:
    60×
  • Description:
    Fluorite ELWD phase objective
  • Seq:
    15
  • Working distance:
    1,5 - 2,2 mm