ContactUsLinkComponent
Centrifuge tubes, borosilicate glass 3.3, round bottom, without rim, 110 x 56 ± 0.8 mm, wall 3.5 ± 0.3 mm 1 * 10 items
Catalog # HECH91980900010
Centrifuge tubes, borosilicate glass 3.3, round bottom, without rim, 110 x 56 ± 0.8 mm, wall 3.5 ± 0.3 mm 1 * 10 items
Catalog # HECH91980900010
Frequently Bought Together