Orders LinkContactUsLinkComponent
VWR® Basic, Shoe Covers
VWR® Basic, Shoe Covers
40 µm, Red, U*
Catalog # 113-9087
Supplier:   VWR Collection
VWR® Basic, Shoe Covers
Catalog # 113-9087
Supplier:  VWR Collection
VWR® Basic, Shoe Covers
VWR® Basic, Shoe Covers
40 µm, Red, U*
Price
68,60 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Disposable shoe covers made of CPE film.
  • Free from natural rubber latex
  • Available in different colours
L×W: 410×170 mm
Thickness: 25, 40 or 70 µm
* One size fits all
Packaging: 10 pieces per bundle, 10 bundles per bag, 10 bags per case.
Specifications
  • Shoe size:
    U*
  • Colour:
    Red
  • Description:
    40 µm
  • Seq:
    05