Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Polyethersulfone 50 kD Sartocon Slice 50, for single use with 50cm² Filtersurface area 1 * 1 items
SARP3D91465050ELLP
:
Sartorius
Polyethersulfone 50 kD Sartocon Slice 50, for single use with 50cm² Filtersurface area 1 * 1 items
SARP3D91465050ELLP
:
Sartorius
Frequently Bought Together
MicroKros filter module 26cm 100kD mPES 1mm dry 1 * 1 items
: Spectrum Laboratories
Cone folded filter papers, Whatman™
: Whatman products (Cytiva)
Filtration capsules, AcroPak™
: Cytiva (Formerly Pall Lab)
In-Line filters, Polydisc™ HD, Whatman™
: Whatman products (Cytiva)
SILICONE NO 8 STOPPER 9/16 HOLE, 5/PK 1 * 5 items
: Merck
Mesh woven filters, stainless steel, Spectra Mesh®
: Spectrum Laboratories
Mesh woven filters, nylon, Spectra Mesh®
: Spectrum Laboratories