Starter Set B for processing NucleoSpin 8-well strips on the Qiagen BioRobots 9600 / 9604 / 3000 - 1 Column Holder B, 1 Column Holde r D, 6 NucleoSpin Dummy Strips 1 * 1 SET
Catalog # MANA740683
Supplier: MACHEREY-NAGEL
Frequently Bought Together