ContactUsLinkComponent
NucleoBond Buffer Set I for the isolation of low-copy plasmids, cosmids, BACs, PACs, and P1 constructs, only applicable with NucleoB ond PC kits, sufficient for 10 AX 500 columns (Maxi preps), 20 AX 100 columns (Midi preps), or 1 * 1 SET
Catalog # MANA740601
Supplier:  MACHEREY-NAGEL
undefined
NucleoBond Buffer Set I for the isolation of low-copy plasmids, cosmids, BACs, PACs, and P1 constructs, only applicable with NucleoB ond PC kits, sufficient for 10 AX 500 columns (Maxi preps), 20 AX 100 columns (Midi preps), or 1 * 1 SET
Catalog # MANA740601
Supplier:  MACHEREY-NAGEL
Frequently Bought Together