ContactUsLinkComponent
NucleoSpin RNA Set for NucleoZOL (10) 10 preps for extraction of total RNA from NucleoZOL lysates - NucleoSpin RNA Columns, Collecti on Tubes, buffers EQ Excepted Quantities or __UN 1993 Flammable liquid, n.o.s. (1,4-dioxane mixture) 1 * 10 items
  MANA740406.10
undefined
NucleoSpin RNA Set for NucleoZOL (10) 10 preps for extraction of total RNA from NucleoZOL lysates - NucleoSpin RNA Columns, Collecti on Tubes, buffers EQ Excepted Quantities or __UN 1993 Flammable liquid, n.o.s. (1,4-dioxane mixture) 1 * 10 items
  MANA740406.10
Frequently Bought Together