Orders LinkContactUsLinkComponent
Screw caps
65 mm, 1000/1500/2000 ml
  215-4246
 :   Kautex
Screw caps
  215-4246
 :  Kautex
Screw caps
Screw caps
65 mm, 1000/1500/2000 ml
154,00 €
1 * 100 items
LDPE, natural.
  • Good chemical resistance
  • Good thermal resistance
  • Leakproof
  • Thread:
    65 mm
  • Seq:
    06
  • For bottles:
    1000/1500/2000 ml
  • Cat. No.:
    215-4246