Orders LinkContactUsLinkComponent
Screw caps
Screw caps
40 mm, 200/250/300/500 ml
Catalog # 215-4244
Supplier:   Kautex
Screw caps
Catalog # 215-4244
Supplier:  Kautex
Screw caps
Screw caps
40 mm, 200/250/300/500 ml
Price
57,60 €
Log in for availability
1 * 100 items
Product Details & Documents
LDPE, natural.
  • Good chemical resistance
  • Good thermal resistance
  • Leakproof
Specifications
  • Thread:
    40 mm
  • Seq:
    04
  • For bottles:
    200/250/300/500 ml