Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
10 ml Precision FAST qPCR Master Mix for capillary LightCycler 1.0-2.0 premixed with SYBRgreen (10x 1ml) 1 * 1.000 Tests
ZPFASTCLSY10ML
:
PRIMERDESIGN
10 ml Precision FAST qPCR Master Mix for capillary LightCycler 1.0-2.0 premixed with SYBRgreen (10x 1ml) 1 * 1.000 Tests
ZPFASTCLSY10ML
:
PRIMERDESIGN
Frequently Bought Together
VWR® Premium Line, Serological Pipettes
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR®, Transfer Pipettes, Graduated
: VWR Collection