Orders LinkContactUsLinkComponent
TRACKMAN® Connected
TRACKMAN® Connected
TRACKMAN® Connected, EU
Catalog # 613-5996
Supplier:   GILSON
TRACKMAN® Connected
Catalog # 613-5996
Supplier:  GILSON
TRACKMAN® Connected
TRACKMAN® Connected
TRACKMAN® Connected, EU
Price
2 031,00 €
Log in for availability
1 * 1 items
Product Details & Documents
TRACKMAN® Connected is a tablet with exclusive accessories and applications that make pipetting on 96- and 384-well plates faster and more reliable, improving your efficiency at the bench by tracking your pipetting tasks.
  • Add traceability in your experiments
  • Boost your experiment reliability
  • Get immediate reproducibility
  • Access pipette analytics and calibration history
Designed to communicate with PIPETMAN® M Connected via Bluetooth®, the tablet interacts in real-time with the pipette and guides you through your protocol with PipettePilot™. When complete, results can be saved, shared, and exported to sciNote, an open-source electronic lab notebook.
Easily capture notes, photos, and monitor your protocols. Streamline your work with the Bluetooth-connected environment sensor by automatically registering a protocol run action such as pipetting and conditions like temperature, humidity, and air pressure.
When paired with PIPETMAN M Connected, the installed PipettePilot app guides you through your microplate method. The app displays where, when, and how much to pipette with real-time interactions to prevent errors.
Effortlessly create and share your protocols. Compatibility with sciNote lets you structure your work into projects, experiments, and tasks, making it easy to visualise your work progress. Sync your results for a comprehensive view of your pipetting actions, samples, and protocols.
PipetteScope™ informs you exactly when your pipette needs to be calibrated. Through the app, you can easily request a calibration appointment and also view service history reports in one central location. PipetteScope gives you instant access to the pipette usage analytics as well as access to the user guide.
Specifications
  • Description:
    TRACKMAN® Connected, EU