Accessories
15 ml holder (adapter and blocking plate) for 6×15 ml lysis tubes, Precellys® Evolution
Catalog # 432-0308
Supplier:   Bertin Instruments
Accessories
Catalog # 432-0308
Supplier:  Bertin Instruments
Accessories
Accessories
15 ml holder (adapter and blocking plate) for 6×15 ml lysis tubes, Precellys® Evolution
Price
1 060,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    15 ml holder (adapter and blocking plate) for 6×15 ml lysis tubes
  • Seq:
    04
  • For:
    Precellys® Evolution