Agilent J&W DB-FATWAX Ultra Inert GC column, 30 m, 0.32 mm, 0.25 µm 1 * 1 items
Catalog # HEWLG3903-63009
Supplier: Agilent
Agilent J&W DB-FATWAX Ultra Inert GC column, 30 m, 0.32 mm, 0.25 µm 1 * 1 items
Catalog # HEWLG3903-63009
Supplier: Agilent
Frequently Bought Together
Safety canisters
Supplier: S.C.A.T.
Certified headspace crimp top vials
Supplier: Agilent
VWR®, Vial Racks
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Capillary columns, CP-FFAP CB
Supplier: VARIAN