RINDLEDERHANDSCHUH VS53 GR 10.5 EN388 3143, EN407 413X4X Länge: 35 cm, Farbe: grau 1 * 12 PAAR
Catalog # FELD0261/10.5
Supplier: FELDTMANN HELMUT
RINDLEDERHANDSCHUH VS53 GR 10.5 EN388 3143, EN407 413X4X Länge: 35 cm, Farbe: grau 1 * 12 PAAR
Catalog # FELD0261/10.5
Supplier: FELDTMANN HELMUT
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection