ContactUsLinkComponent
Amarell-Special-Therm., +48+102:0,1C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid , c.450x9,0-9,5mm, Richter-top finish, with ice point scale, null
Catalog # AMARN61622-CB
Supplier:  Amarell
Amarell-Special-Therm., +48+102:0,1C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid , c.450x9,0-9,5mm, Richter-top finish, with ice point scale, null
Catalog # AMARN61622-CB
Supplier:  Amarell
Frequently Bought Together