ContactUsLinkComponent
Gallium-Quartz-glass therm., +500+1000:5°C, 400x8-9mm, cap. colourless, strip bearing the scale: quartz, immersion 150mm, pin-top fi nish with button, suitable for government verification 1 * 1 items
Catalog # AMARQ140054-GA
Supplier:  Amarell
Gallium-Quartz-glass therm., +500+1000:5°C, 400x8-9mm, cap. colourless, strip bearing the scale: quartz, immersion 150mm, pin-top fi nish with button, suitable for government verification 1 * 1 items
Catalog # AMARQ140054-GA
Supplier:  Amarell
Frequently Bought Together